missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13176-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Can specifically activate hydrolysis of GTP bound to RAC1 and CDC42, but not RALA. Mediates ATP-dependent transport of S-(2,4-dinitrophenyl)-glutathione (DNP-SG) and doxorubicin (DOX) and is the major ATP-dependent transporter of glutathione conjugates of electrophiles (GS-E) and DOX in erythrocytes. Can catalyze transport of glutathione conjugates and xenobiotics, and may contribute to the multidrug resistance phenomenon. Serves as a scaffold protein that brings together proteins forming an endocytotic complex during interphase and also with CDC2 to switch off endocytosis, One of its substrates would be EPN1/Epsin.Specifications
RALBP1 | |
Polyclonal | |
Unconjugated | |
RALBP1 | |
76 kDa Ral interacting protein, DNP SG ATPase, Ral interacting protein 1, ralA binding protein 1, RALBP1, RIP1, RLIP1, RLIP76 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10928 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15311 | |
RALBP1 | |
RALBP1 Fusion Protein Ag3843 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |