missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGPS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RALGPS1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881534
|
Proteintech
13368-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871534
|
Proteintech
13368-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RALGPS1 gene ontology annotations related to this gene include regulation of Ral protein signal transduction; small GTPase mediated signal transduction.Specifications
RALGPS1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
A2AR50, Q5JS13 | |
241308, 9649 | |
RALGPS1 Fusion Protein Ag4187 | |
Primary | |
-20°C | |
RALGPS1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA0351, RalA exchange factor RalGPS1, RalGEF 2, RALGEF2, RALGPS1, RALGPS1A | |
RALGPS1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RALGPS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech