Learn More
RANTES Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12000-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879443
  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
The RANTES antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RANTES. This antibody recognizes human antigen. The RANTES antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RANTES
Polyclonal
Unconjugated
20 μL
6352
IgG
ELISA, Neutralization, Western Blot
207 μg/mL