Learn More
RAP1GDS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10377-1-AP-20UL
 View more versions of this product
Product Code. 16886723
  • 1645.54 SEK / 20µL
Description
The smg GDP dissociation stimulator (smgGDS) protein is a stimulatory GDP/GTP exchange protein with GTPase activity (Riess et al., 1993).
Specifications
Specifications
RAP1GDS1
Polyclonal
Unconjugated
RAP1GDS1
Exchange factor smgGDS, GDS1, RAP1GDS1, SMG GDS protein, SmgGDS
Rabbit
Antigen Affinity Chromatography
RUO
229877, 5910
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P52306
RAP1GDS1
RAP1GDS1 Fusion Protein Ag0230
20 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RAP1GDS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated