Learn More
RASEF Mouse anti-Human, Unconjugated, Clone: 1C2B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67159-1-IG-150UL
Description
The RASEF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RASEF. This antibody recognizes human antigen. The RASEF antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RASEF | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1C2B2 | |
Unconjugated | |
150 μL | |
158158 | |
IgG2a |