Learn More
RASEF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11569-1-AP-150UL
 View more versions of this product
Product Code. 16888733
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Binds predomitly GDP, and also GTP.
Specifications
Specifications
RASEF
Polyclonal
Unconjugated
RASEF
RAB45, Ras related protein Rab 45, RASEF
Rabbit
Antigen Affinity Chromatography
RUO
158158, 242505
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5RI75, Q8IZ41
RASEF
RASEF Fusion Protein Ag2149
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RASEF Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated