missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RASSF6 |
---|---|
Concentration | 0.53 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879303
|
Proteintech
11921-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869303
|
Proteintech
11921-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.Specifications
RASSF6 | |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q4QR82, Q6ZTQ3, Q80UQ2 | |
166824, 305251, 73246 | |
RASSF6 Fusion Protein Ag2565 | |
Primary | |
-20°C | |
RASSF6 |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RASSF6 | |
RASSF6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RASSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech