Learn More
Novus Biologicals™ Rat BNP-32 ELISA Kit (Colorimetric)
specific to Brain Natriuretic Peptide 32
Brand:  Novus Biologicals™ NBP3-43779
Product Code. 30281808
  • 6391.84 SEK / 1 set
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.031-2 ng/ml (example only; lot dependent)
Colorimetric
This Rat BNP ELISA Kit (Colorimetric) is specific to Brain Natriuretic Peptide 32 (103-134).
ELISA, Sandwich ELISA
NPPB
Storage of components varies. See protocol for specific instructions.
0.025 ng/ml (example only; lot dependent)
Serum, Plasma, Cell Culture, Tissue Extracts
Rat
4879
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Rat BNP-32 ELISA Kit (Colorimetric) > 1 Kit