Learn More
Novus Biologicals™ Rat BNP-32 ELISA Kit (Colorimetric)
specific to Brain Natriuretic Peptide 32
Brand: Novus Biologicals™ NBP3-43779
This item is not returnable.
View return policy
Description
Specifications
0.031-2 ng/ml (example only; lot dependent) | |
Colorimetric | |
This Rat BNP ELISA Kit (Colorimetric) is specific to Brain Natriuretic Peptide 32 (103-134). | |
ELISA, Sandwich ELISA | |
NPPB | |
Storage of components varies. See protocol for specific instructions. |
0.025 ng/ml (example only; lot dependent) | |
Serum, Plasma, Cell Culture, Tissue Extracts | |
Rat | |
4879 | |
1 Kit |
Product Content Correction
Product Title
Novus Biologicals™ Rat BNP-32 ELISA Kit (Colorimetric) > 1 Kit
Spot an opportunity for improvement?Share a Content Correction