Learn More
Novus Biologicals™ Rat Galanin R2/GALR2 ELISA Kit (Colorimetric)
Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: Novus Biologicals™ NBP2-76731
This item is not returnable.
View return policy
Description
The Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) measures rat Galanin R2/GALR2 in serum, plasma and other biological fluids.
Specifications
1.25 to 80 ng/mL | |
ELISA Kit (Colorimetric) | |
8811 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.75 ng/mL | |
Biological Fluids, Plasma, Serum | |
GALR2 | |
1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) > 1 Kit
Spot an opportunity for improvement?Share a Content Correction