Learn More
Novus Biologicals™ Rat Galanin R2/GALR2 ELISA Kit (Colorimetric)
Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  Novus Biologicals™ NBP2-76731
Product Code. 18323321
  • 7940.00 SEK / 1 set
Description
The Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) measures rat Galanin R2/GALR2 in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.25 to 80 ng/mL
ELISA Kit (Colorimetric)
8811
Rat
Storage of components varies. See protocol for specific instructions.
0.75 ng/mL
Biological Fluids, Plasma, Serum
GALR2
1 Kit
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Rat Galanin R2/GALR2 ELISA Kit (Colorimetric) > 1 Kit