Learn More
Novus Biologicals™ Rat GPT ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-18697
This item is not returnable.
View return policy
Description
Specifications
0.5 - 32 ng/mL (example only; lot dependent) | |
ELISA Kit (Colorimetric) | |
ELISA | |
GPT | |
metabolism | |
AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1 |
0.27 ng/mL (example only; lot dependent) | |
Cell Culture, Plasma, Serum, Tissue Extracts, Urine | |
2875 | |
1 Kit | |
Storage is content dependent. | |
Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Rat GPT ELISA Kit (Colorimetric) > 1 Kit
Spot an opportunity for improvement?Share a Content Correction