missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Ig2A, Isotype Control, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems IC006V
This item is not returnable.
View return policy
Description
IgG2a Monoclonal specifically detects IgG2a in Rat samples. It is validated for Control.
Specifications
IgG2a | |
54447 | |
Control 5 uL/10^6 cells | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Selected for use as rat IgG 2A isotype control for flow cytometry applications. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
KLH | |
200 Tests | |
Secondary | |
Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rat Ig2A, Isotype Control, R&D Systems™ > 200 Tests; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction