missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Rat Recombinant GM-CSF

Click to view available options
Quantity:
100 μg
1000 μg
20 μg
Unit Size:
1000µg
100µg
20µg
Description
- Granulocyte-macrophage colony-stimulating factor (GM-CSF) promotes the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages
- In synergy with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, it also stimulates erythroid and megakaryocyte progenitor cells
- GM-CSF is produced by multiple cell types, including stromal cells, Paneth cells, macrophages, dendritic cells (DCs), endothelial cells, smooth muscle cells, fibroblasts, chondrocytes, and Th1 and Th17 cells T cells
- The receptor for GM-CSF (GM-CSFR) is composed of two subunits: the cytokine-specific α subunit (GMRα; CD116) and the common subunit βc (CD131) shared with IL-3 and IL-5 receptors
Specifications
Specifications
Name | Colony-stimulating factor 2, CSF-2, MGI-1GM, Pluripoietic-alpha |
Quantity | 20 μg |
Cell Type | Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Monocytes; Myeloid Cells |
Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Rat Recombinant GM-CSF
Spot an opportunity for improvement?Share a Content Correction