Learn More
Novus Biologicals™ Rat SCF/c-kit Ligand ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-06958
This item is not returnable.
View return policy
Description
Specifications
15.63 to 1000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat SCF/c-kit Ligand in samples. No significant cross-reactivity or interference between Rat SCF/c-kit Ligand and analogues was observed. | |
4254 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Rat |
9.38 pg/mL | |
Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
KITLG | |
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Rat SCF/c-kit Ligand ELISA Kit (Colorimetric) > 1 Kit
Spot an opportunity for improvement?Share a Content Correction