Learn More
Novus Biologicals™ Rat SCF/c-kit Ligand ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand:  Novus Biologicals™ NBP3-06958
Product Code. 18332493
  • 7900.00 SEK / 1 set
Specifications
15.63 to 1000 pg/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat SCF/c-kit Ligand in samples. No significant cross-reactivity or interference between Rat SCF/c-kit Ligand and analogues was observed.
4254
1 Kit
Storage of components varies. See protocol for specific instructions.
Rat
9.38 pg/mL
Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
KITLG
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Rat SCF/c-kit Ligand ELISA Kit (Colorimetric) > 1 Kit