missing translation for 'onlineSavingsMsg'
Learn More

RB1 Mouse anti-Human, Unconjugated, Clone: 1A2A6, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen RB1
Clone 1A2A6
Concentration 1000 ÎĽg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16880196
View Documents
Proteintech
67521-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16870196
View Documents
Proteintech
67521-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RB1. This antibody recognizes human antigen. The RB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

RB1
1000 ÎĽg/mL
Monoclonal
Mouse
5925
Primary
1A2A6
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Human
IgG2a
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RB1 Mouse anti-Human, Unconjugated, Clone: 1A2A6, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde ĂĽbermittelt