missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25628-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ZNF639 binds DNA and may function as a transcriptional repressor.Specifications
RB1 | |
Polyclonal | |
Unconjugated | |
RB1 | |
OSRC, p105 Rb, pp110, pRb, RB, RB1, retinoblastoma 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5925 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P06400 | |
RB1 | |
RB1 Fusion Protein Ag22578 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |