missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | RB1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884195
|
Proteintech
25628-1-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874195
|
Proteintech
25628-1-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF639 binds DNA and may function as a transcriptional repressor.Specifications
RB1 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P06400 | |
5925 | |
RB1 Fusion Protein Ag22578 | |
Primary | |
-20°C | |
RB1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
OSRC, p105 Rb, pp110, pRb, RB, RB1, retinoblastoma 1 | |
RB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |