missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10043-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The immunogen of this antibody is C-terminal 154aa of RB1CC1.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
RB1CC1 | |
Polyclonal | |
Unconjugated | |
RB1CC1 | |
RB1CC1 | |
Rabbit | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TDY2, Q9ESK9 | |
RB1CC1 | |
Antigen Affinity Chromatography | |
RUO | |
12421, 312927, 9821 | |
-20°C | |
Liquid |