missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBAP48 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20364-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to RBBP4.
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation.Specifications
RBAP48 | |
Polyclonal | |
Unconjugated | |
RBBP4 | |
CAF 1 subunit C, CAF I 48 kDa subunit, CAF I p48, Histone binding protein RBBP4, NURF55, RBAP48, RBBP 4, RBBP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19646, 5928 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q09028, Q60972 | |
RBBP4 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RBAP48 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction