missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13181-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.Specifications
RBBP5 | |
Polyclonal | |
Unconjugated | |
RBBP5 | |
RBBP 5, RBBP5, RBQ3, SWD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213464, 304794, 5929 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15291, Q8BX09 | |
Rbbp5 | |
RBBP5 Fusion Protein Ag3848 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |