missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18893-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Regulates the p53 pathway to control the expansion growth of digestive organs.Specifications
RBM16 | |
Polyclonal | |
Unconjugated | |
SCAF8 | |
CCAP7, KIAA1116, RBM16, RNA binding motif protein 16, RNA binding protein 16, SCAF8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
106583, 22828 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6DID3, Q9UPN6 | |
Scaf8 | |
RBM16 Fusion Protein Ag13499 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |