missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RBM16 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878724
|
Proteintech
18893-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868724
|
Proteintech
18893-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Regulates the p53 pathway to control the expansion growth of digestive organs.Specifications
RBM16 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6DID3, Q9UPN6 | |
106583, 22828 | |
RBM16 Fusion Protein Ag13499 | |
Primary | |
-20°C | |
SCAF8 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CCAP7, KIAA1116, RBM16, RNA binding motif protein 16, RNA binding protein 16, SCAF8 | |
Scaf8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |