Learn More
RBM24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18178-1-AP-20UL
 View more versions of this product
Product Code. 16898234
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
RBM24 is preferentially expressed in muscle during differentiation in vitro and may regulate myogenic differentiation.
Specifications
Specifications
RBM24
Polyclonal
Unconjugated
RBM24
RBM24, RNA binding motif protein 24, RNA binding protein 24, RNPC6
Rabbit
Antigen Affinity Chromatography
RUO
221662, 666794, 690139
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
D3Z4I3, Q9BX46
Rbm24
RBM24 Fusion Protein Ag12871
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RBM24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated