Learn More
RBM39 Mouse anti-Human, Unconjugated, Clone: 2D2C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67420-1-IG-150UL
Description
The RBM39 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBM39. This antibody recognizes human, mouse, rat antigen. The RBM39 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
RBM39 | |
Monoclonal | |
Unconjugated | |
150 μL | |
9584 | |
IgG2b |
Western Blot, Immunohistochemistry, Immunofluorescence | |
2D2C8 | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.