Learn More
RBM39 Mouse anti-Human, Unconjugated, Clone: 2D2C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67420-1-IG-150UL
 View more versions of this product
Product Code. 16890036
  • 4652.90 SEK / 150µL
Description
The RBM39 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBM39. This antibody recognizes human, mouse, rat antigen. The RBM39 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
RBM39
Monoclonal
Unconjugated
150 μL
9584
IgG2b
Western Blot, Immunohistochemistry, Immunofluorescence
2D2C8
Mouse
Primary
Human, Rat, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RBM39 Mouse anti-Human, Unconjugated, Clone: 2D2C8, Proteintech > 150 μL