missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15784-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May bind RNA.Specifications
RBM41 | |
Polyclonal | |
Unconjugated | |
RBM41 | |
bB383K5.1, RBM41, RNA binding motif protein 41, RNA binding protein 41 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
237073, 55285, 680581 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8JZV4, Q96IZ5 | |
RBM41 | |
RBM41 Fusion Protein Ag8483 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |