Learn More
RBM46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24368-1-AP-20UL
This item is not returnable.
View return policy
Description
RBM46 is a protein with RNA binding function.Specifications
RBM46 | |
Polyclonal | |
Unconjugated | |
RBM46 | |
Cancer/testis antigen 68, CT68, RBM46, RNA binding motif protein 46 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
166863 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TBY0 | |
RBM46 | |
RBM46 Fusion Protein Ag21653 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
RBM46 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction