missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBMY1A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14239-1-AP-150UL
This item is not returnable.
View return policy
Description
RBMY1A1 is a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. This protein is thought to function as a splicing regulator during spermatogenesis. Multiple closely related paralogs of this gene are found in a gene cluster in the AZFb azoospermia factor region of chromosome Y. Most of these related copies are thought to be pseudogenes, though several likely encode functional proteins.Specifications
RBMY1A1 | |
Polyclonal | |
Unconjugated | |
RBMY1A1 | |
hRBMY, RBM1, RBM2, RBMY, RBMY1A1, RBMY1C, RNA binding motif protein 1, RNA binding motif protein 2, YRRM1, YRRM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5940 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0DJD3 | |
RBMY1A1 | |
RBMY1A1 Fusion Protein Ag5493 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RBMY1A1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction