Learn More
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
Name | Elongation factor Tu Protein |
Quantity | 500 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | tuf1 |
Protein Length | Full Length |
Product Type | Recombinant Protein |
Cross Reactivity | A. baumannii |
Species | Yeast |
Protein Tag | His |
Show More |
Product Content Correction
Product Title
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein >
Spot an opportunity for improvement?Share a Content Correction