missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacillus Unsaturated glucuronyl hydrolase Protein
A DNA sequence encoding the Bacillus sp. (strain GL1) Unsaturated glucuronyl hydrolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8079-ec-50ug
This item is not returnable.
View return policy
Specifications
Unsaturated glucuronyl hydrolase Protein | |
Research Use Only | |
ugl | |
Recombinant Protein | |
E. coli | |
MWQQAIGDAL GITARNLKKF GDRFPHVSDG SNKYVLNDNT DWTDGFWSGI LWLCYEYTGD EQYREGAVRT VASFRERLDR FENLDHHDIG FLYSLSAKAQ WIVEKDESAR KLALDAADVL MRRWRADAGI IQAWGPKGDP ENGGRIIIDC LLNLPLLLWA GEQTGDPEYR RVAEAHALKS RRFLVRGDDS SYHTFYFDPE NGNAIRGGTH QGNTDGSTWT RGQAWGIYGF ALNSRYLGNA DLLETAKRMA RHFLARVPED GVVYWDFEVP QEPSSYRDSS ASAITACGLL EIASQLDESD PERQRFIDAA KTTVTALRDG YAERDDGEAE GFIRRGSYHV RGGISPDDYT IWGDYYYLEA LLRLERGVTG YWYERGR | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Bacillus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction