missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Fibritin Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Fibritin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7036-10ug
This item is not returnable.
View return policy
Specifications
NP_049771.1 | |
Fibritin Protein | |
Research Use Only | |
wac | |
Recombinant Protein | |
E. coli | |
TDIVLNDLPF VDGPPAEGQS RISWIKNGEE ILGADTQYGS EGSMNRPTVS VLRNVEVLDK NIGILKTSLE TANSDIKTIQ GILDVSGDIE ALAQIGINKK DISDLKTLTS EHTEILNGTN NTVDSILADI GPFNAEANSV YRTIRNDLLW IKRELGQYTG QDINGLPVVG NPSSGMKHRI INNTDVITSQ GIRLSELETK FIESDVGSLT IEVGNLREEL GPKPPSFSQN VYSRLNEIDT KQTTVESDIS AIKTSIGYPG NNSIITSVNT NTDNIASINL ELNQSGGIKQ RLTVIETSIG SDDIPSSIKG QIKDNTTSIE SLNGIVGENT SSGLRANVSW LNQIVGTDSS GGQPSPPGSL LNRVSTIETS VSGLNNAVQN LQVEIGNNSA GIKGQVVALN TLVNGTNPNG STVEERGLTN SIKANETNIA SVTQEVNTAK GNISSLQGDV QALQEAGYIP EAPRDGQAYV RKDGEWVFLS TFLSPA | |
Greater than 90% as determined by SDS-PAGE. |
1258630 | |
10 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Bacteriophage Fibritin Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction