missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine CCL20 / MIP-3 alpha Protein

A DNA sequence encoding the Bos taurus (Bovine) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7906-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041521

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_776688.1
CCL20 / MIP-3 alpha Protein
Research Use Only
CCL20
Recombinant Protein
Yeast
ASNFDCCLRY TERILHPSIL VGFTQQLANE ACDINAVVFY TRKKLAVCAD PKKKWVKQVV HMLSQRVKRM
Greater than 90% as determined by SDS-PAGE.
281666
10 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions