missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Chicken Riboflavin-binding Protein Product Code.: 16056831

enQuireBio™ Recombinant Chicken Riboflavin-binding Protein

Product Code. 16056831
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16056831

Brand: enQuireBio™ QP8851ye500ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Gallus gallus (Chicken) Riboflavin-binding protein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_990794.1
Gene ID (Entrez) 396449
Name Riboflavin-binding Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RBP
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Chicken
Species Yeast
Protein Tag His
Sequence QQYGCLEGDT HKANPSPEPN MHECTLYSES SCCYANFTEQ LAHSPIIKVS NSYWNRCGQL SKSCEDFTKK IECFYRCSPH AARWIDPRYT AAIQSVPLCQ SFCDDWYEAC KDDSICAHNW LTDWERDESG ENHCKSKCVP YSEMYANGTD MCQSMWGESF KVSESSCLCL QMNKKDMVAI KHLLSESSEE SSSMSSSEEH ACQKKLLK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Chicken Riboflavin-binding Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.