Learn More
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein Product Code.: 15934008
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein
Product Code. 15934008
100 μg, 100µg
Click to view available options
Quantity:
100 μg
250 μg
Unit Size:
100µg
250µg
Product Code. 15934008
Brand: enQuireBio™ QP10484100ug
Recombinant Protein
Specifications
Name Serotype 4 VLP Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Untested
Product Type Recombinant Protein
Cross Reactivity Dengue virus
Species Human
Protein Tag Native
Buffer 10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.
Purity or Quality Grade >95% as determined by SDS-PAGE.
Product Content Correction
Product Title
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein >