missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S21 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S21, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_417537.1 |
---|---|
Gene ID (Entrez) | 947577 |
Name | 30S ribosomal protein S21 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025591
|
enQuireBio™
QP8616-ec-10ug |
10 ÎĽg | N/A | ||||||
16065591
|
enQuireBio™
QP8616-ec-50ug |
50 ÎĽg | N/A | ||||||
16015591
|
enQuireBio™
QP8616-ec-100ug |
100 ÎĽg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16045591
|
enQuireBio™
QP8616-ec-200ug |
200 ÎĽg | N/A | ||||||
16055591
|
enQuireBio™
QP8616-ec-500ug |
500 ÎĽg | N/A | ||||||
16035591
|
enQuireBio™
QP8616-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_417537.1 | |
30S ribosomal protein S21 Protein | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |
947577 | |
Research Use Only | |
rpsU | |
Recombinant Protein | |
E. coli | |
PVIKVRENEP FDVALRRFKR SCEKAGVLAE VRRREFYEKP TTERKRAKAS AVKRHAKKLA RENARRTRLY | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli 30S ribosomal protein S21 Protein