missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Bifunctional protein HldE Protein
A DNA sequence encoding the Escherichia coli (strain K12) Bifunctional protein HldE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6949-200ug
This item is not returnable.
View return policy
Specifications
NP_417524.1 | |
Bifunctional protein HldE Protein | |
Research Use Only | |
hldE | |
Recombinant Protein | |
E. coli | |
MKVTLPEFER AGVMVVGDVM LDRYWYGPTS RISPEAPVPV VKVNTIEERP GGAANVAMNI ASLGANARLV GLTGIDDAAR ALSKSLADVN VKCDFVSVPT HPTITKLRVL SRNQQLIRLD FEEGFEGVDP QPLHERINQA LSSIGALVLS DYAKGALASV QQMIQLARKA GVPVLIDPKG TDFERYRGAT LLTPNLSEFE AVVGKCKTEE EIVERGMKLI ADYELSALLV TRSEQGMSLL QPGKAPLHMP TQAQEVYDVT GAGDTVIGVL AATLAAGNSL EEACFFANAA AGVVVGKLGT STVSPIELEN AVRGRADTGF GVMTEEELKL AVAAARKRGE KVVMTNGVFD ILHAGHVSYL ANARKLGDRL IVAVNSDAST KRLKGDSRPV NPLEQRMIVL GALEAVDWVV SFEEDTPQRL IAGILPDLLV KGGDYKPEEI AGSKEVWANG GEVLVLNFED GCSTTNIIKK IQQDKKG | |
Greater than 90% as determined by SDS-PAGE. |
947548 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Bifunctional protein HldE Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction