missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein

Product Code. 16057470
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001033658.1
Name CFA / I fimbrial subunit E Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol cfaE
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species Yeast
Protein Tag His
Sequence MNKILFIFTL FFSSGFFTFA VSADKNPGSE NMTNTIGPHD RGGSSPIYNI LNSYLTAYNG SHHLYDRMSF LCLSSQNTLN GACPSSDAPG TATIDGETNI TLQFTEKRSL IKRELQIKGY KQFLFKNANC PSKLALNSSH FQCNREQASG ATLSLYIPAG ELNKLPFGGV WNAVLKLNVK RRYDTTYGTY TINITVNLTD KGNIQIWLPQ FKSNARVDLN LRPTGGGTYI GRNSVDMCFY DGYSTNSSSL EIRFQDDNSK SDGKFYLKKI NDDSKELVYT LSLLLAGKNL TPTNGQALNI NTASLETNWN RITAVTMPEI SVPVLCWPGR LQLDAKVKNP EAGQYMGNIK ITFTPSSQTL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.