missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cyclophilin A / PPIA / CYPA Protein

A DNA sequence encoding the Escherichia coli O157:H7 Cyclophilin A / PPIA / CYPA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7335-ec-10ug

 View more versions of this product

Product Code. 16018290

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_312241.1
Cyclophilin A / PPIA / CYPA Protein
Research Use Only
PPIA
Recombinant Protein
E. coli
AKGDPHVLLT TSAGNIELEL DKQKAPVSVQ NFVDYVNSGF YNNTTFHRVI PGFMIQGGGF TEQMQQKKPN PPIKNEADNG LRNTRGTIAM ARTADKDSAT SQFFINVADN AFLDHGQRDF GYAVFGKVVK GMDVADKISQ VPTHDVGPYQ NVPSKPVVIL SAKVLP
Greater than 90% as determined by SDS-PAGE.
915930
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Cyclophilin A / PPIA / CYPA Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.