missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant E. coli Inositol-1-monophosphatase Protein Product Code.: 16015551

enQuireBio™ Recombinant E. coli Inositol-1-monophosphatase Protein

Product Code. 16015551
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16015551

Brand: enQuireBio™ QP8609ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Inositol-1-monophosphatase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_417028.1
Gene ID (Entrez) 947285
Name Inositol-1-monophosphatase Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ssyA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag GST
Sequence MHPMLNIAVR AARKAGNLIA KNYETPDAVE ASQKGSNDFV TNVDKAAEAV IIDTIRKSYP QHTIITEESG ELEGTDQDVQ WVIDPLDGTT NFIKRLPHFA VSIAVRIKGR TEVAVVYDPM RNELFTATRG QGAQLNGYRL RGSTARDLDG TILATGFPFK AKQYATTYIN IVGKLFNECA DFRRTGSAAL DLAYVAAGRV DGFFEIGLRP WDFAAGELLV REAGGIVSDF TGGHNYMLTG NIVAGNPRVV KAMLANMRDE LSDALKR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Inositol-1-monophosphatase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.