missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein II Protein
A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein II, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7296-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_416400.1 | |
Methyl-accepting chemotaxis protein II Protein | |
Research Use Only | |
tar | |
Recombinant Protein | |
E. coli | |
IRRMLLTPLA KIIAHIREIA GGNLANTLTI DGRSEMGDLA QSVSHMQRSL TDTVTHVREG SDAIYAGTRE IAAGNTDLSS RTEQQASALE ETAASMEQLT ATVKQNADNA RQASQLAQSA SDTAQHGGKV VDGVVKTMHE IADSSKKIAD IISVIDGIAF QTNILALNAA VEAARAGEQG RGFAVVAGEV RNLASRSAQA AKEIKALIED SVSRVDTGSV LVESAGETMN NIVNAVTRVT DIMGEIASAS DEQSRGIDQV ALAVSEMDRV TQQNASLVQE SAAAAAALEE QASRLTQAVS AFRLAASPLT NKPQTPSRPA SEQPPAQPRL RIAEQDPNWE TF | |
Greater than 90% as determined by SDS-PAGE. |
946399 | |
500 ÎĽg | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein II Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction