missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli nrdD Protein

Product Code. 16036811
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) nrdD, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_418659.1
Gene ID (Entrez) 948755
Name nrdD Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol nrdD
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence MTPHVMKRDG CKVPFKSERI KEAILRAAKA AEVDDADYCA TVAAVVSEQM QGRNQVDINE IQTAVENQLM SGPYKQLARA YIEYRHDRDI EREKRGRLNQ EIRGLVEQTN ASLLNENANK DSKVIPTQRD LLAGIVAKHY ARQHLLPRDV VQAHERGDIH YHDLDYSPFF PMFNCMLIDL KGMLTQGFKM GNAEIEPPKS ISTATAVTAQ IIAQVASHIY GGTTINRIDE VLAPFVTASY NKHRKTAEEW NIPDAEGYAN SRTIKECYDA FQSLEYEVNT LHTANGQTPF VTFGFGLGTS WESRLIQESI LRNRIAGLGK NRKTAVFPKL VFAIRDGLNH KKGDPNYDIK QLALECASKR MYPDILNYDQ VVKVTGSFKT PMGCRSFLGV WENENGEQIH DGRNNLGVIS LNLPRIALEA KGDEATFWKL LDERLVLARK ALMTRIARLE GVKARVAPIL YMEGACGVRL NADDDVSEIF KNGRASISLG YIGIHETINA LFGGEHVYDN EQLRAKGIAI VERLRQAVDQ WKEETGYGFS LYSTPSENLC DRFCRLDTAE FGVVPGVTDK GYYTNSFHLD VEKKVNPYDK IDFEAPYPPL ANGGFICYGE YPNIQHNLKA LEDVWDYSYQ HVPYYGTNTP IDECYECGFT GEFECTSKGF TCPKCGNHDA SRVSVTRRVC GYLGSPDARP FNAGKQEEVK RRVKHLGNGQ IG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli nrdD Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.