missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ornithine decarboxylase, inducible Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ornithine decarboxylase, inducible, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_415220.1 |
---|---|
Gene ID (Entrez) | 945297 |
Name | Ornithine decarboxylase, inducible Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006971
|
enQuireBio™
QP8883-ec-10ug |
10 ÎĽg | N/A | ||||||
16046971
|
enQuireBio™
QP8883-ec-50ug |
50 ÎĽg | N/A | ||||||
16096961
|
enQuireBio™
QP8883-ec-100ug |
100 ÎĽg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16026971
|
enQuireBio™
QP8883-ec-200ug |
200 ÎĽg | N/A | ||||||
16036971
|
enQuireBio™
QP8883-ec-500ug |
500 ÎĽg | N/A | ||||||
16016971
|
enQuireBio™
QP8883-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_415220.1 | |
Ornithine decarboxylase, inducible Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945297 | |
Research Use Only | |
speF | |
Recombinant Protein | |
E. coli | |
MSKLKIAVSD SCPDCFTTQR ECIYINESRN IDVAAIVLSL NDVTCGKLDE IDATGYGIPV FIATENQERV PAEYLPRISG VFENCESRRE FYGRQLETAA SHYETQLRPP FFRALVDYVN QGNSAFDCPG HQGGEFFRRH PAGNQFVEYF GEALFRADLC NADVAMGDLL IHEGAPCIAQ QHAAKVFNAD KTYFVLNGTS SSNKVVLNAL LTPGDLVLFD RNNHKSNHHG ALLQAGATPV YLETARNPYG FIGGIDAHCF EESYLRELIA EVAPQRAKEA RPFRLAVIQL GTYDGTIYNA RQVVDKIGHL CDYILFDSAW VGYEQFIPMM ADCSPLLLDL NENDPGILVT QSVHKQQAGF SQTSQIHKKD SHIKGQQRYV PHKRMNNAFM MHASTSPFYP LFAALNINAK MHEGVSGRNM WMDCVVNGIN ARKLILDNCQ HIRPFVPELV DGKPWQSYET AQIAVDLRFF QFVPGEHWHS FEGYAENQYF VDPCKLLLTT PGIDARNGEY EAFGVPATIL ANFLRENGVV PEKCDLNSIL FLLTPAEDMA KLQQLVALLV RFEKLLESDA PLAEVLPSIY KQHEERYAGY TLRQLCQEMH DLYARHNVKQ LQKEMFRKEH FPRVSMNPQE ANYAYLRGEV ELVRLPDAEG RIAAEGALPY PPGVLCVVPG EIWGGAVLRY FSALEEGINL LPGFAPELQG VYIEEHDGRK QVWCYVIKPR DAQSTLLKGE KL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Ornithine decarboxylase, inducible Protein