missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) Outer-membrane lipoprotein carrier protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7432-ec-1mg
This item is not returnable.
View return policy
Specifications
WP_001295343.1 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Outer-membrane lipoprotein carrier Protein | |
Research Use Only | |
lolA | |
Recombinant Protein | |
E. coli | |
DAASDLKSRL DKVSSFHASF TQKVTDGSGA AVQEGQGDLW VKRPNLFNWH MTQPDESILV SDGKTLWFYN PFVEQATATW LKDATGNTPF MLIARNQSSD WQQYNIKQNG DDFVLTPKAS NGNLKQFTIN VGRDGTIHQF SAVEQDDQRS SYQLKSQQNG AVDAAKFTFT PPQGVTVDDQ RK | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction