missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Purine nucleoside phosphorylase DeoD-type Protein
A DNA sequence encoding the Escherichia coli (strain K12) Purine nucleoside phosphorylase DeoD-type, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8935-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418801.1 | |
Purine nucleoside phosphorylase DeoD-type Protein | |
Research Use Only | |
deoD | |
Recombinant Protein | |
E. coli | |
ATPHINAEMG DFADVVLMPG DPLRAKYIAE TFLEDAREVN NVRGMLGFTG TYKGRKISVM GHGMGIPSCS IYTKELITDF GVKKIIRVGS CGAVLPHVKL RDVVIGMGAC TDSKVNRIRF KDHDFAAIAD FDMVRNAVDA AKALGIDARV GNLFSADLFY SPDGEMFDVM EKYGILGVEM EAAGIYGVAA EFGAKALTIC TVSDHIRTHE QTTAAERQTT FNDMIKIALE SVLLGDKE | |
Greater than 90% as determined by SDS-PAGE. |
945654 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |