missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Purine nucleoside phosphorylase DeoD-type Protein
A DNA sequence encoding the Escherichia coli (strain K12) Purine nucleoside phosphorylase DeoD-type, was expressed in the hosts and tags indicated.
3070.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_418801.1 |
---|---|
Gene ID (Entrez) | 945654 |
Name | Purine nucleoside phosphorylase DeoD-type Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047231
|
enQuireBio™
QP8935-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087231
|
enQuireBio™
QP8935-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037231
|
enQuireBio™
QP8935-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067231
|
enQuireBio™
QP8935-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077231
|
enQuireBio™
QP8935-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057231
|
enQuireBio™
QP8935-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_418801.1 | |
Purine nucleoside phosphorylase DeoD-type Protein | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |
945654 | |
Research Use Only | |
deoD | |
Recombinant Protein | |
E. coli | |
ATPHINAEMG DFADVVLMPG DPLRAKYIAE TFLEDAREVN NVRGMLGFTG TYKGRKISVM GHGMGIPSCS IYTKELITDF GVKKIIRVGS CGAVLPHVKL RDVVIGMGAC TDSKVNRIRF KDHDFAAIAD FDMVRNAVDA AKALGIDARV GNLFSADLFY SPDGEMFDVM EKYGILGVEM EAAGIYGVAA EFGAKALTIC TVSDHIRTHE QTTAAERQTT FNDMIKIALE SVLLGDKE | |
Greater than 90% as determined by SDS-PAGE. |