missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein

A DNA sequence encoding the Escherichia coli (strain K12) RNA-splicing ligase RtcB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7212-ye-100ug

 View more versions of this product

Product Code. 16017740

  • 12935.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_417879.1
RNA-splicing ligase RtcB Protein
Research Use Only
rtcB
Recombinant Protein
Yeast
MNYELLTTEN APVKMWTKGV PVEADARQQL INTAKMPFIF KHIAVMPDVH LGKGSTIGSV IPTKGAIIPA AVGVDIGCGM NALRTALTAE DLPENLAELR QAIETAVPHG RTTGRCKRDK GAWENPPVNV DAKWAELEAG YQWLTQKYPR FLNTNNYKHL GTLGTGNHFI EICLDESDQV WIMLHSGSRG IGNAIGTYFI DLAQKEMQET LETLPSRDLA YFMEGTEYFD DYLKAVAWAQ LFASLNRDAM MENVVTALQS ITQKTVRQPQ TLAMEEINCH HNYVQKEQHF GEEIYVTRKG AVSARAGQYG IIPGSMGAKS FIVRGLGNEE SFCSCSHGAG RVMSRTKAKK LFSVEDQIRA TAHVECRKDA EVIDEIPMAY KDIDAVMAAQ SDLVEVIYTL RQVVCVKG
Greater than 81% as determined by SDS-PAGE.
947929
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.