missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Trigger factor Protein

A DNA sequence encoding the Escherichia coli O127:H6 (strain E2348/69 / EPEC) Trigger factor, was expressed in the hosts and tags indicated.

3070.00 SEK - 40175.00 SEK

Specifications

Accession Number WP_001198386.1
Name Trigger factor Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol tig
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16098870
View Documents
enQuireBio™
QP7443-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16038880
View Documents
enQuireBio™
QP7443-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16088870
View Documents
enQuireBio™
QP7443-ec-100ug
100 ÎĽg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16018880
View Documents
enQuireBio™
QP7443-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028880
View Documents
enQuireBio™
QP7443-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008880
View Documents
enQuireBio™
QP7443-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

WP_001198386.1
Research Use Only
tig
Recombinant Protein
E. coli
MQVSVETTQG LGRRVTITIA ADSIETAVKS ELVNVAKKVR IDGFRKGKVP MNIVAQRYGA SVRQDVLGDL MSRNFIDAII KEKINPAGAP TYVPGEYKLG EDFTYSVEFE VYPEVELQGL EAIEVEKPIV EVTDADVDGM LDTLRKQQAT WKEKDGAVEA EDRVTIDFTG SVDGEEFEGG KASDFVLAMG QGRMIPGFED GIKGHKAGEE FTIDVTFPEE YHAENLKGKA AKFAINLKKV EERELPELTA EFIKRFGVED GSVEGLRAEV RKNMERELKS AIRNRVKSQA IEGLVKANDI DVPAALIDSE IDVLRRQAAQ RFGGNEKQAL ELPRELFEEQ AKRRVVVGLL LGEVIRTNEL KADEERVKGL IEEMASAYED PKEVIEFYSK NKELMDNMRN VALEEQAVEA VLAKAKVTEK ETTFNELMNQ QA
Greater than 90% as determined by SDS-PAGE.
Trigger factor Protein
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Trigger factor Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.