missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C Virus polyProtein

A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C Virus polyprotein, was expressed in the hosts and tags indicated.

4340.00 SEK - 26260.00 SEK

Specifications

Name Hepatitis C Virus polyProtein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16027251
View Documents
enQuireBio™
QP8939-ec-10ug
10 ÎĽg
4740.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
16067251
View Documents
enQuireBio™
QP8939-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16017251
View Documents
enQuireBio™
QP8939-ec-100ug
100 ÎĽg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16047251
View Documents
enQuireBio™
QP8939-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16057251
View Documents
enQuireBio™
QP8939-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16037251
View Documents
enQuireBio™
QP8939-ec-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Hepatitis C Virus polyProtein
Not determined.
Recombinant Protein
E. coli
APITAYAQQT RGLLSTIVTS LTGRDTNENC GEVQVLSTAT QSFLGTAVNG VMWTVYHGAG SKTISGPQGP VNQMYTNVDQ DLVGWPAPPG VRSLAPCTCG ASDLYLVTRH ADVVPVRRRG DTRGALLSPR PISTLKGSSG GPLLCPMGHA AGIFRAAVCT RGVAKAVDFV PVESLETTMR SPVFTDNSTP PAVPQTYQVA HLHAPTGSGK STKVPAAYAA QGYKVLVLNP SVAATLGFGA YMSKAYGIDP NIRTGVRTIT TGAPITYSTY GKFLADGGCS GGAYDIIICD ECHSTDATTI LGIGTALDQA ETAGMRLVVL ATATPPGSVT TPHSNIEEVA LPTTGEVPFY GRAIPLGLIK GGRHLIFCHS KKKCDELAGQ LTSLGLNAVA YYRGLDVSVI PTSGDVVVCA TDALMTGFTG DFDSVIDCNT SVIQTIDFSL DPTFSIETTT VPQDAVSRSQ RRGRTGRGRL GIYRYVTPGE RPSGIFDTAV LCECYDAGCA WYELTPAETT VRLRAYFNTP GLPVCQDHLE FWESVFTGLT NIDGHFLSQT KQSGENFPYL VAYQATVCAR ALAPPPSWDT IWKCLTRLKP TLHGPTPLLY RLGSVQNEVT LTHPITKYIM ACMSADLEVV T
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Partial (see sequence information for more details).
Hepatitis
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Hepatitis C Virus polyProtein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.