missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Hepatitis C Virus polyProtein Product Code.: 16027251

enQuireBio™ Recombinant Hepatitis C Virus polyProtein

Product Code. 16027251
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16027251

Brand: enQuireBio™ QP8939ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C Virus polyprotein, was expressed in the hosts and tags indicated.

Specifications

Name Hepatitis C Virus polyProtein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Hepatitis
Species E. coli
Protein Tag His
Sequence APITAYAQQT RGLLSTIVTS LTGRDTNENC GEVQVLSTAT QSFLGTAVNG VMWTVYHGAG SKTISGPQGP VNQMYTNVDQ DLVGWPAPPG VRSLAPCTCG ASDLYLVTRH ADVVPVRRRG DTRGALLSPR PISTLKGSSG GPLLCPMGHA AGIFRAAVCT RGVAKAVDFV PVESLETTMR SPVFTDNSTP PAVPQTYQVA HLHAPTGSGK STKVPAAYAA QGYKVLVLNP SVAATLGFGA YMSKAYGIDP NIRTGVRTIT TGAPITYSTY GKFLADGGCS GGAYDIIICD ECHSTDATTI LGIGTALDQA ETAGMRLVVL ATATPPGSVT TPHSNIEEVA LPTTGEVPFY GRAIPLGLIK GGRHLIFCHS KKKCDELAGQ LTSLGLNAVA YYRGLDVSVI PTSGDVVVCA TDALMTGFTG DFDSVIDCNT SVIQTIDFSL DPTFSIETTT VPQDAVSRSQ RRGRTGRGRL GIYRYVTPGE RPSGIFDTAV LCECYDAGCA WYELTPAETT VRLRAYFNTP GLPVCQDHLE FWESVFTGLT NIDGHFLSQT KQSGENFPYL VAYQATVCAR ALAPPPSWDT IWKCLTRLKP TLHGPTPLLY RLGSVQNEVT LTHPITKYIM ACMSADLEVV T
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Hepatitis C Virus polyProtein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.