missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-related protein 2/3 complex subunit 3, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_001265485.1 |
---|---|
Gene ID (Entrez) | 10094 |
Name | Actin-related protein 2 / 3 complex subunit 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094621
|
enQuireBio™
QP8439-ec-10ug |
10 ÎĽg | N/A | ||||||
16034631
|
enQuireBio™
QP8439-ec-50ug |
50 ÎĽg | N/A | ||||||
16084621
|
enQuireBio™
QP8439-ec-100ug |
100 ÎĽg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16014631
|
enQuireBio™
QP8439-ec-200ug |
200 ÎĽg | N/A | ||||||
16024631
|
enQuireBio™
QP8439-ec-500ug |
500 ÎĽg | N/A | ||||||
16004631
|
enQuireBio™
QP8439-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001265485.1 | |
Actin-related protein 2 / 3 complex subunit 3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10094 | |
Research Use Only | |
ARPC3 | |
Recombinant Protein | |
E. coli | |
PAYHSSLMDP DTKLIGNMAL LPIRSQFKGP APRETKDTDI VDEAIYYFKA NVFFKNYEIK NEADRTLIYI TLYISECLKK LQKCNSKSQG EKEMYTLGIT NFPIPGEPGF PLNAIYAKPA NKQEDEVMRA YLQQLRQETG LRLCEKVFDP QNDKPSKWWT CFVKRQFMNK SLSG | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 3 Protein