missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Adamts7 Protein

A DNA sequence encoding the Homo sapiens (Human) Adamts7, was expressed in the hosts and tags indicated.

1640.00 SEK - 22640.00 SEK

Specifications

Accession Number NP_055087.2
Gene ID (Entrez) 11173
Name Adamts7 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16003531
View Documents Promotion Details
enQuireBio™
QP8246-EC-100UG
100 μg
4960.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16023531
View Documents Promotion Details
enQuireBio™
QP8246-EC-1MG
1 mg
22640.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16013531
View Documents Promotion Details
enQuireBio™
QP8246-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16033531
View Documents Promotion Details
enQuireBio™
QP8246-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16043531
View Documents Promotion Details
enQuireBio™
QP8246-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16053531
View Documents Promotion Details
enQuireBio™
QP8246-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_055087.2
Adamts7 Protein
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
11173
Research Use Only
ADAMTS7
Recombinant Protein
E. coli
KWVETLVVAD AKMVEYHGQP QVESYVLTIM NMVAGLFHDP SIGNPIHITI VRLVLLEDEE EDLKITHHAD NTLKSFCKWQ KSINMKGDAH PLHHDTAILL TRKDLCAAMN RPCETLGLSH VAGMCQPHRS CSINEDTGLP LAFTVAHELG HSFGIQHDGS GNDCEPVGKR PFIMSPQLLY DAAPLTWSRC SRQYITRFLD RGWGLCLDDP PAKDIIDFPS VPPGVLYDVS HQCRLQYGAY SAFCEDMDNV CHTLWCSVGT TCHSKLDAAV DGTRCGENKW CLSGECVPVG FRPEAVDGGW SGWSAWSICS RSCGMGVQSA ERQCTQPTPK YKGRYCVGER KRFRLCNLQA CP
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications